Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine 95%

Supplier: Thermo Fisher Scientific

H52115.MD H52115.03
ALFAH52115.MDEA 228 SGD
ALFAH52115.MD ALFAH52115.03
Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine 95%
Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine

Purity: 95% CAS No.: 401915-53-5 Molecular Formula: C35H42N4O7S Formula Weight: 662.80 MDL No.: MFCD03788172

Formula: C₃₅H₄₂N₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD03788172
CAS Number: 401915-53-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR