2-Benzyl-N-boc-D-proline 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Boc-D-α-Bzl-Pro-OH, (S)-2-Benzyl-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid

ALFAH52186.MDEA 0 SGD
ALFAH52186.MD
2-Benzyl-N-boc-D-proline 95%
2-Benzyl-N-boc-D-proline

Purity: 95% CAS No.: 706806-61-3 Molecular Formula: C17H23NO4 Formula Weight: 305.37 Melting Point: 146° MDL No.: MFCD06659153

Formula: C₁₇H₂₃NO₄
MW: 305.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659153
CAS Number: 706806-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR