4-Bromo-2-fluorobenzamide 96%

Supplier: Thermo Fisher Scientific
Warning

H52336.MD H52336.03
ALFAH52336.MDEA 155 SGD
ALFAH52336.MD ALFAH52336.03
4-Bromo-2-fluorobenzamide 96%
4-Bromo-2-fluorobenzamide

Purity: 96% CAS No.: 292621-45-5 Molecular Formula: C7H5BrFNO Formula Weight: 218.03 Melting Point: 151-153° MDL No.: MFCD03094432

Formula: C₇H₅BrFNO
MW: 218.03 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD03094432
CAS Number: 292621-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR