6-Bromo-1-tetralone 96%

Supplier: Thermo Fisher Scientific

Synonyms: 6-Bromo-3,4-dihydro-1(2H)-naphthalenone, 6-Bromo-3,4-dihydronaphthalen-1(2H)-one

H52381.03
ALFAH52381.03EA 760 SGD
ALFAH52381.03
6-Bromo-1-tetralone 96%
6-Bromo-1-tetralone

Purity: 96% CAS No.: 66361-67-9 Molecular Formula: C10H9BrO Formula Weight: 225.09 Storage Temperature: 2°C to 8°C Melting Point: 40-42° MDL No.: MFCD04114378

Formula: C₁₀H₉BrO
Melting Pt: 40…42 °C
MDL Number: MFCD04114378
CAS Number: 66361-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR