2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Chloro-5-(methoxycarbonyl)phenylboronic acid, (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid, 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

ALFAH52999.MDEA 0 SGD
ALFAH52999.MD
2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥98%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid

Purity: 98% CAS No.: 913835-92-4 Molecular Formula: C8H8BClO4 Formula Weight: 214.41 Melting Point: 230-232° MDL No.: MFCD08689528

Formula: C₈H₈BClO₄
MW: 214.41 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD08689528
CAS Number: 913835-92-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR