Ethyl-3-(trimethylsilyl)propiolate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 3-(trimethylsilyl)propynoate, 3-(Trimethylsilyl)-2-propynoic acid ethyl ester, 3-(Trimethylsilyl)propiolic acid ethyl ester, Ethyl 3-(trimethylsilyl)propiolate

H53517.06 H53517.14
ALFAH53517.06EA 132 SGD
ALFAH53517.06 ALFAH53517.14
Ethyl-3-(trimethylsilyl)propiolate ≥98%
Ethyl 3-(trimethylsilyl)propiolate

Purity: 98% CAS No.: 16205-84-8 Molecular Formula: C8H14O2Si Formula Weight: 170.28 Flash Point: 71°(160°F) Refractive Index: 1.441 Boiling Point: 92-96°/28mm Beilstein Reference: 1767850 MDL No.: MFCD00074886

Formula: C₈H₁₄O₂Si
MW: 170.28 g/mol
Boiling Pt: 92…96 °C (28 mmHg)
Density: 0.897
Flash Pt: 71 °C (160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074886
CAS Number: 16205-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR