2-(4-Nitrophenyl)ethanol ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol

H54504.03 H54504.06 H54504.14
ALFAH54504.03EA 40.6 SGD
ALFAH54504.03 ALFAH54504.06 ALFAH54504.14
2-(4-Nitrophenyl)ethanol ≥98%
2-(4-Nitrophenyl)ethanol

Purity: 98% CAS No.: 100-27-6 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 59-62° Boiling Point: 177°/16mm MDL No.: MFCD00010202

Formula: C₈H₉NO₃
Boiling Pt: 177 °C (16 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00010202
CAS Number: 100-27-6
EINECS: 202-835-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR