2-Bromo-3-hexylthiophene 98%

Supplier: Thermo Fisher Scientific
Warning

H56243.06
ALFAH56243.06EA 410 SGD
ALFAH56243.06
2-Bromo-3-hexylthiophene 98%
2-Bromo-3-hexylthiophene

Purity: 98% CAS No.: 69249-61-2 Molecular Formula: C10H15BrS Formula Weight: 247.20 Flash Point: 110°(230°F) Refractive Index: 1.529 MDL No.: MFCD09907959

Formula: C₁₀H₁₅BrS
MW: 247.20 g/mol
Density: 1.240
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD09907959
CAS Number: 69249-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR