Azetidine ≥98%

Supplier: Thermo Scientific
Danger

H56351.03
ALFAH56351.03EA 339 SGD
ALFAH56351.03
Azetidine ≥98%
Azetidine
Formula: C₃H₇N
MW: 57,1 g/mol
Boiling Pt: 61…62 °C
Density: 0,847
Flash Pt: –21 °C (–6 °F)
MDL Number: MFCD00005165
CAS Number: 503-29-7
EINECS: 207-963-8
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR