4-Bromo-1-indanone 97%

Supplier: Thermo Fisher Scientific
Warning

H56937.06 H56937.09
ALFAH56937.06EA 282 SGD
ALFAH56937.06 ALFAH56937.09
4-Bromo-1-indanone 97%
4-Bromo-1-indanone

Purity: 97% CAS No.: 15115-60-3 Molecular Formula: C9H7BrO Formula Weight: 211.06 Melting Point: 95-99° MDL No.: MFCD01719772

Formula: C₉H₇BrO
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01719772
CAS Number: 15115-60-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR