(R)-Pyrrolidin-3-ylmethanol 95%

Supplier: Thermo Fisher Scientific
Warning

H57871.MD H57871.03
ALFAH57871.MDEA 230 SGD
ALFAH57871.MD ALFAH57871.03
(R)-Pyrrolidin-3-ylmethanol 95%
(R)-Pyrrolidin-3-ylmethanol

Purity: 95% CAS No.: 110013-18-8 Molecular Formula: C5H11NO Formula Weight: 101.15 Storage Temperature: 2°C to 8°C Boiling Point: 173-177° MDL No.: MFCD09607969

Formula: C₅H₁₁NO
Boiling Pt: 173…177 °C
Density: 0,979
MDL Number: MFCD09607969
CAS Number: 110013-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR