N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-[(tert-Butoxycarbonyl)amino]hexanoic Acid, 6-(Boc-amino)hexanoic Acid, Boc-6-Ahx-OH, N-Boc-6-aminohexanoic Acid, Boc-ε-Acp-OH

H61350.06 H61350.14
ALFAH61350.06EA 0 SGD
ALFAH61350.06 ALFAH61350.14
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 95%
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid

Purity: 95% CAS No.: 6404-29-1 Molecular Formula: C11H21NO4 Formula Weight: 231.29 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 35-40° Beilstein Reference: 2049561 MDL No.: MFCD00037798

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Melting Pt: 35…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037798
CAS Number: 6404-29-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR