(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%

Supplier: Thermo Fisher Scientific

Synonyms: Boc-Lys(Boc)-OH, Nα,Nepsilon-Di-Boc-L-lysine

H62019.22 H62019.14
ALFAH62019.22EA 0 SGD
ALFAH62019.22 ALFAH62019.14
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid

Purity: 98% CAS No.: 2483-46-7 Molecular Formula: C16H30N2O6 Formula Weight: 346.42 Storage Temperature: -30°C to -10°C Physical Form: Low melting solid Appearance: White MDL No.: MFCD00038515

Formula: C₁₆H₃₀N₂O₆
MW: 346.4 g/mol
MDL Number: MFCD00038515
CAS Number: 2483-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR