9-Bromo-1-nonene 97%

Supplier: Thermo Fisher Scientific
Warning

H64792.03 H64792.06
ALFAH64792.03EA 0 SGD
ALFAH64792.03 ALFAH64792.06
9-Bromo-1-nonene 97%
9-Bromo-1-nonene

Purity: 97% CAS No.: 89359-54-6 Molecular Formula: C9H17Br Formula Weight: 205.14 Physical Form: Liquid Refractive Index: 1.4675 Appearance: Colourless to pale yellow Boiling Point: 100-101°/15mm Beilstein Reference: 1746338 MDL No.: MFCD09037826

Formula: C₉H₁₇Br
MW: 205.139 g/mol
Boiling Pt: 100…101 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD09037826
CAS Number: 89359-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR