8-Bromo-1-octene 97%

Supplier: Thermo Fisher Scientific
Warning

H64830.03 H64830.06
ALFAH64830.03EA 0 SGD
ALFAH64830.03 ALFAH64830.06
8-Bromo-1-octene 97%
8-Bromo-1-octene

Purity: 97% CAS No.: 2695-48-9 Molecular Formula: C8H15Br Formula Weight: 191.11 Flash Point: 78°(172°F) Physical Form: Liquid Density: 1.139 Refractive Index: 1.4670 Appearance: Pale yelllow Boiling Point: 92°/24mm Beilstein Reference: 2234481 MDL No.: MFCD00000275

Formula: C₈H₁₅Br
Boiling Pt: 92 °C (24 mmHg)
Density: 1,139
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000275
CAS Number: 2695-48-9
EINECS: 220-268-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR