Casein
Supplier: Thermo Fisher Scientific
J12840.Q1
ALFAJ12840.Q1EA 0 SGD
ALFAJ12840.Q1
Casein
Proteins and Peptides
Hammarsten casein is solvent extracted to remove fats from whole casein by the procedure of Hammarsten.
  • Protein: ≥95%
  • Loss on drying: ≤10,0%