Yeast extract, powder, ultrapure

Supplier: Thermo Fisher Scientific

J23547.A1
ALFAJ23547.A1EA 128 SGD
ALFAJ23547.A1
Yeast extract, powder, ultrapure
Media for Microbiology Dehydrated Media for Microbiology

  • Assay Protein (TN×6.25): 57 - 72%
  • Assay Salmonella: Negative
  • Assay Standard Plate Count: ≤15000/g
  • Residual Water(at Time of Packaging): ≤6%

Order Now

SPECIFICATIONS

Environmentally Preferable
Storage conditions Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR