Polyvinylpyrrolidone, M.W. ∼ 10,000 (K15)

Supplier: Thermo Fisher Scientific

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

J60382.14 J60382.22 J60382.36
ALFAJ60382.14EA 59.5 SGD
ALFAJ60382.14 ALFAJ60382.22 ALFAJ60382.36
Polyvinylpyrrolidone, M.W. ∼ 10,000 (K15)
Polyvinylpyrrolidone

CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,7697 MDL No.: MFCD00149016


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR