Water DNase-, RNase-free

Supplier: Thermo Fisher Scientific

J60610.XC
ALFAJ60610.XCEA 248 SGD
ALFAJ60610.XC
Water DNase-, RNase-free
Water

High quality.


  • Used in all molecular biology applications
  • Suitable for all RNA work
  • No toxic agents in purification procedures


RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work. Toxic agents such as DEPC are not used in purification procedures.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1,000
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR