Magnesium sulphate 1 M in aqueous solution, sterile-filtered

Supplier: Thermo Fisher Scientific

Synonyms: MgSO4, Magnesium sulphate anhydrous

J61030.XE J61030.AK
ALFAJ61030.XEEA 0 SGD
ALFAJ61030.XE ALFAJ61030.AK
Magnesium sulphate 1 M in aqueous solution, sterile-filtered
Magnesium sulphate

CAS No.: 7487-88-9
Molecular Formula: MgO4S
Formula Weight: 120.37
Physical Form: Liquid
MDL No.: MFCD00011110

Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR