TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt) ≥98%

Supplier: Thermo Fisher Scientific
Warning

J61189.14 J61189.22
ALFAJ61189.14EA 0 SGD
ALFAJ61189.14 ALFAJ61189.22
TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt) ≥98%
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)

Purity: 98% CAS No.: 91000-53-2 Molecular Formula: C7H16NNaO6S Formula Weight: 265.26 Physical Form: Powder Appearance: White MDL No.: MFCD00069903


Zwitterionic buffer

Formula: C₇H₁₇NO₆S
Storage Temperature: Ambient
MDL Number: MFCD00069903
CAS Number: 91000-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR