Sodium alginate, high viscosity

Supplier: Thermo Fisher Scientific

Synonyms: Alginic acid sodium salt, Algin

J61887.22 J61887.30 J61887.A1
ALFAJ61887.22EA 0 SGD
ALFAJ61887.22 ALFAJ61887.30 ALFAJ61887.A1
Sodium alginate, high viscosity
Sodium alginate

CAS No.: 9005-38-3 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Merck Reference: 14,241 MDL No.: MFCD00081310


A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR