Lithium chloride 1 M in aqueous solution

Supplier: Thermo Fisher Scientific

ALFAJ61895.AEEA 0 SGD
ALFAJ61895.AE ALFAJ61895.AD
Lithium chloride 1 M in aqueous solution
Lithium chloride

CAS No.: 7447-41-8 Molecular Formula: ClLi Formula Weight: 42.39 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00011078

Formula: LiCl
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR