Brefeldin A ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

J62340.MB J62340.MF
ALFAJ62340.MBEA 0 SGD
ALFAJ62340.MB ALFAJ62340.MF
Brefeldin A ≥99%
Brefeldin A
Formula: C₁₆H₂₄O₄
Melting Pt: 201…205 °C
Storage Temperature: Freezer
MDL Number: MFCD00083258
CAS Number: 20350-15-6
Merck Index: 14,1369

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR