Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30)

Supplier: Thermo Fisher Scientific

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

J62417.36 J62417.A1 J62417.22
ALFAJ62417.36EA 0 SGD
ALFAJ62417.36 ALFAJ62417.A1 ALFAJ62417.22
Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30)
Polyvinylpyrrolidone

CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,7697 MDL No.: MFCD00149016


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR