Staurosporine ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

ALFAJ62837.MAEA 0 SGD
ALFAJ62837.MA
Staurosporine ≥99%
Staurosporine

Purity: 99+% CAS No.: 62996-74-1 Molecular Formula: C28H26N4O3 Formula Weight: 466.54 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Pale yellow Melting Point: 240-243° Solubility: Soluble in DMSO or ethanol Beilstein Reference: 1060573 MDL No.: MFCD00077402


Applications: Broad spectrum protein kinase inhibitor

Formula: C₂₈H₂₆N₄O₃
Melting Pt: 240…243 °C
MDL Number: MFCD18252446
CAS Number: 62996-74-1
Merck Index: 14,8802

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR