BCA protein assay kit

Supplier: Thermo Fisher Scientific

ALFAJ63283.QAEA 0 SGD
ALFAJ63283.QA
BCA protein assay kit
Assays Cellular Assays

BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. The kit accurately measures protein concentrations from 0,5 to 30 μg/ml in tube assays and 1 to 20 μg/ml in 96- or 384-well plate assays. It is used for studying protein:protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.


  • Clear liquid


Physical Form: Liquid Appearance: Clear liquid


Applications: Contains two separate solutions for estimation of protein concentration.


Delivery information: Contains 250 ml of Solution A (Bicinchonic acid and tartrate in an alkaline carbonate buffer) and 5,5 ml of Solution B (4% w/v cupric sulfate).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR