Guanidinium thiocyanate for molecular biology

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

J65104.18 J65104.30 J65104.36
ALFAJ65104.18EA 0 SGD
ALFAJ65104.18 ALFAJ65104.30 ALFAJ65104.36
Guanidinium thiocyanate for molecular biology
Guanidinium thiocyanate

Guanidine thiocyanate is used for RNA isolation and can solubilise proteins. When the compound is used to solubilize proteins it can be removed by dialysis against urea without difficulty. The hybridization of guanidine thiocyanate-solubilize hepatocytes can be used as a method to quantify mRNA from hepatocytes. Blood sample lysate stored in guanidine thiocyanate has been shown to maintain nucleic acid integrity. Research describes guanidine thiocyanate to be ineffective for RNA isolation of plant tissue with a high levels of secondary metabolites and storage proteins.


Soluble in H₂O (1420 g/L) at 20°C, and ethanol (5%).


Caution: Light Sensitive. Keep container tightly closed and store away from oxidising agents.

Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118…122 °C
Density: 1.29
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Specification Test Results

Appearance (Colour) White to off-white
Form Crystalline powder
Assay (Titration) 99.0% min.
DNase None detected
Protease activity None detected
RNase None detected
UV/Vis A280 (70%, water) 0.8
UV/Vis A300 (6M, water) 0.1
UV/Vis A410 (6M, water) 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR