Agarose Separation ≥ 500 bp, Genetic Performance Certified, Ultrapure, Molecular Biology Grade

Supplier: Thermo Fisher Scientific

J75817.30
ALFAJ75817.30EA 0 SGD
ALFAJ75817.30
Agarose Separation ≥ 500 bp, Genetic Performance Certified, Ultrapure, Molecular Biology Grade
Agarose

Recommended for PCR, DNA/RNA analytical separation, preparative DNA/RNA recovery, DNA/RNA in-gel recovery, and blotting.

MDL Number: MFCD00081294
CAS Number: 9012-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR