Guanidinium hydrochloride 99.5+% for molecular biology

Supplier: Thermo Fisher Scientific
Warning

J75823.36
ALFAJ75823.36EA 484 SGD
ALFAJ75823.36
Guanidinium hydrochloride 99.5+% for molecular biology
Guanidinium hydrochloride

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies

Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185…188 °C
Density: 1,344
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 14,04562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form White/clear crystals
Assay, Absorbance (225 nm, 6 M, 1 cm) ≤0.2
Assay, Absorbance (260 nm, 6 M, 1 cm) ≤0.03
Assay, Aldehyde None detected
Assay, Ammeline None detected
Assay ≥99.5%
Impurity content, Sulfate (SO₄) ≤ 50 ppm
Melting Point 184 ±3°C
Solution Test, (6 M, H₂O) Clear and colourless
Identification (FTIR) Conforms
Dnase (endo) Passes Test
Protease Activity Passes Test
RNase Passes Test
Clear and colorless ≤1 ppm
Trace Metal: Arsenic (As) Solution Test (6M, H2O)
Trace Metal: Copper (Cu) ≤1 ppm
Trace Metal: Iron (Fe) ≤2 ppm
Trace Metal: Lead (Pb) ≤5 ppm
Trace Metal: Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR