TBS with Tween® (TBST), 20X solution, Molecular Biology Grade, ultrapure

Supplier: Thermo Scientific

J77500.K8
ALFAJ77500.K8EA 914 SGD
ALFAJ77500.K8
TBS with Tween® (TBST), 20X solution, Molecular Biology Grade, ultrapure
Buffers Electrophoresis Buffers Washing Buffers

TBS is used in Western blotting protocols.


  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • Protease: Passes test


Solution is 0,2 μm filtered and is supplied in an upright carton with a convenient dispensing spout.


Delivery information: 20X TBS (Tris Buffered Saline) solution consisting of 500 mM Tris, 60 mM KCl, 2,8 M NaCl, and 1,0% Tween 20 in high purity dH₂O adjusted to pH 7,4.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR