Ethyl indole-2-carboxylate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

L00797.06 L00797.14
ALFAL00797.06EA 112 SGD
ALFAL00797.06 ALFAL00797.14
Ethyl indole-2-carboxylate ≥98%
Ethyl indole-2-carboxylate

Purity: 98+% CAS No.: 3770-50-1 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 122-125° Beilstein Reference: 146395

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR