4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥99%

Supplier: Thermo Fisher Scientific
Warning

L00982.06 L00982.14
ALFAL00982.06EA 132 SGD
ALFAL00982.06 ALFAL00982.14
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥99%
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole

Purity: 99+% CAS No.: 1750-12-5 Molecular Formula: C2H6N6S Formula Weight: 146.17 Melting Point: ca 220° dec. Beilstein Reference: 134867 UN No.: 1325 MDL No.: MFCD00003098

Formula: C₂H₆N₆S
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003098
CAS Number: 1750-12-5
EINECS: 217-135-8
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR