1,3-Bis(trimethylsilyl)urea 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N,N'-Bis(trimethylsilyl)urea

L01301.18 L01301.30
ALFAL01301.18EA 70.2 SGD
ALFAL01301.18 ALFAL01301.30
1,3-Bis(trimethylsilyl)urea 98
1,3-Bis(trimethylsilyl)urea

Purity: 98+% CAS No.: 18297-63-7 Molecular Formula: C7H20N2OSi2 Formula Weight: 204.42 Melting Point: ca 230° dec. Beilstein Reference: 1937452

Formula: C₇H₂₀N₂OSi₂
MW: 204.42 g/mol
Melting Pt: ∼ 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008260
CAS Number: 18297-63-7
EINECS: 242-177-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR