1-Naphthalenemethanol ≥98%

Supplier: Thermo Fisher Scientific

L01347.09 L01347.18
ALFAL01347.09EA 88.5 SGD
ALFAL01347.09 ALFAL01347.18
1-Naphthalenemethanol ≥98%
1-Naphthalenemethanol

Purity: 98+% CAS No.: 4780-79-4 Molecular Formula: C11H10O Formula Weight: 158.20 Melting Point: 58-63° Boiling Point: 301°/715mm MDL No.: MFCD00004044

Formula: C₁₁H₁₀O
Boiling Pt: 301 °C (715 mmHg)
Melting Pt: 58…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00004044
CAS Number: 4780-79-4
EINECS: 225-324-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR