3-Bromo-1,2,4,5-tetramethylbenzene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-2,3,5,6-tetramethylbenzene

L01465.06 L01465.14
ALFAL01465.06EA 173 SGD
ALFAL01465.06 ALFAL01465.14
3-Bromo-1,2,4,5-tetramethylbenzene ≥99%
3-Bromo-1,2,4,5-tetramethylbenzene

Purity: 99% CAS No.: 1646-53-3 Molecular Formula: C10H13Br Formula Weight: 213.12 Melting Point: 58-62° Boiling Point: 115-120°/5mm MDL No.: MFCD00000070

Formula: C₁₀H₁₃Br
Boiling Pt: 115…120 °C (5 mmHg)
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000070
CAS Number: 1646-53-3
EINECS: 216-707-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR