Ethyl cyclohexanecarboxylate 98

Supplier: Thermo Fisher Scientific

L02622.06 L02622.14
ALFAL02622.06EA 179 SGD
ALFAL02622.06 ALFAL02622.14
Ethyl cyclohexanecarboxylate 98
Ethyl cyclohexanecarboxylate

Purity: 98+% CAS No.: 3289-28-9 Molecular Formula: C9H16O2 Formula Weight: 156.23 Refractive Index: 1.4420 Boiling Point: 194-196° MDL No.: MFCD00041734

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Boiling Pt: 194…196 °C
Density: 0.936
Storage Temperature: Ambient
MDL Number: MFCD00041734
CAS Number: 3289-28-9
EINECS: 221-945-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR