(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97%

Supplier: Thermo Fisher Scientific
Warning

L02814.22 L02814.36 L02814.0E
ALFAL02814.22EA 71.5 SGD
ALFAL02814.22 ALFAL02814.36 ALFAL02814.0E
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Purity: 97% CAS No.: 100-79-8 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 80°(176°F) Refractive Index: 1.4340 Melting Point: -27° Boiling Point: 188-190° Beilstein Reference: 104465 Merck Reference: 14,5213 MDL No.: MFCD00063238

Formula: C₆H₁₂O₃
Boiling Pt: 188…190 °C
Melting Pt: –27 °C
Density: 1,064
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063238
CAS Number: 100-79-8
EINECS: 202-888-7
Merck Index: 14,5213

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR