4-Cyclohexylaniline ≥97%

Supplier: Thermo Fisher Scientific
Warning

L03184.03
ALFAL03184.03EA 131 SGD
ALFAL03184.03
4-Cyclohexylaniline ≥97%
4-Cyclohexylaniline

Purity: 97% CAS No.: 6373-50-8 Molecular Formula: C12H17N Formula Weight: 175.28 Melting Point: 52-56° Boiling Point: 166°/13mm MDL No.: MFCD00001454

Formula: C₁₂H₁₇N
MW: 175.27 g/mol
Boiling Pt: 166 °C (13 mmHg)
Melting Pt: 52…56 °C
Density: 1.13 g/cm³ (–100.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001454
CAS Number: 6373-50-8
EINECS: 228-918-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR