Ethyl-2-methylnicotinate 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 2-methylnicotinate

L03256.06 L03256.14
ALFAL03256.06EA 164 SGD
ALFAL03256.06 ALFAL03256.14
Ethyl-2-methylnicotinate 97
Ethyl 2-methylnicotinate

Purity: 97% CAS No.: 1721-26-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 102°(215°F) Refractive Index: 1.5050 Odour: Recognizable Boiling Point: 107-108°/13mm Beilstein Reference: 118200 MDL No.: MFCD00006336

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 107…108 °C (13 mmHg)
Density: 1.070
Flash Pt: 102 °C (215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006336
CAS Number: 1721-26-2
EINECS: 217-013-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR