Benzyl alcohol 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

L03292.30 L03292.0B L03292.0E
ALFAL03292.30EA 71 SGD
ALFAL03292.30 ALFAL03292.0B ALFAL03292.0E
Benzyl alcohol 99
Benzyl alcohol

Purity: 99% CAS No.: 100-51-6 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 100°(212°F) Refractive Index: 1.5400 Odour: Aromatic Melting Point: -15° Boiling Point: 205° Beilstein Reference: 878307 Merck Reference: 14,1124 MDL No.: MFCD00004599

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205 °C
Melting Pt: –15 °C
Density: 1.045
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR