1-Bromo-4-cyclohexylbenzene 98

Supplier: Thermo Fisher Scientific

L04636.14 L04636.06
ALFAL04636.14EA 581 SGD
ALFAL04636.14 ALFAL04636.06
1-Bromo-4-cyclohexylbenzene 98
1-Bromo-4-cyclohexylbenzene

Purity: 98% CAS No.: 25109-28-8 Molecular Formula: C12H15Br Formula Weight: 239.16 Refractive Index: 1.5595 Boiling Point: 90-94°/0.01mm MDL No.: MFCD00019339

Formula: C₁₂H₁₅Br
MW: 239.16 g/mol
Boiling Pt: 90…94 °C (0.01 mmHg)
Density: 1.287
Storage Temperature: Ambient
MDL Number: MFCD00019339
CAS Number: 25109-28-8
EINECS: 246-623-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR