(±)-α-Lipoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

L04711.06 L04711.14
ALFAL04711.06EA 156 SGD
ALFAL04711.06 ALFAL04711.14
(±)-α-Lipoic acid ≥98%
(±)-α-Lipoic acid

Purity: 98% CAS No.: 1077-28-7 Molecular Formula: C8H14O2S2 Formula Weight: 206.33 Storage Temperature: 2°C to 8°C Melting Point: 59-63° Boiling Point: 160-165° Beilstein Reference: 81853 Merck Reference: 14,9326 MDL No.: MFCD00005474

Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Boiling Pt: 160…165 °C
Melting Pt: 59…63 °C
MDL Number: MFCD00005474
CAS Number: 1077-28-7
EINECS: 214-071-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR