3-Bromo-5-nitrosalicylaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

L07071.03 L07071.06 L07071.14
ALFAL07071.03EA 72.4 SGD
ALFAL07071.03 ALFAL07071.06 ALFAL07071.14
3-Bromo-5-nitrosalicylaldehyde ≥98%
3-Bromo-5-nitrosalicylaldehyde

Purity: 98% CAS No.: 16789-84-7 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 147-151° Beilstein Reference: 1213613

Formula: C₇H₄BrNO₄
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00051833
CAS Number: 16789-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR