3-Methyl-1-phenyl-2-butanone 96%

Supplier: Thermo Fisher Scientific

L07176.06
ALFAL07176.06EA 213 SGD
ALFAL07176.06
3-Methyl-1-phenyl-2-butanone 96%
3-Methyl-1-phenyl-2-butanone

Purity: 96% CAS No.: 2893-05-2 Molecular Formula: C11H14O Formula Weight: 162.23 Refractive Index: 1.5040 Boiling Point: 237-240° MDL No.: MFCD00040487

Formula: (C₆H₅)CH₂C(O)CH(CH₃)CH₃
Boiling Pt: 237…240 °C
Density: 0,968
Storage Temperature: Ambient
MDL Number: MFCD00040487
CAS Number: 2893-05-2
EINECS: 220-765-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR