2-Bromo-4-nitroanisole ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromo-1-methoxy-4-nitrobenzene

L07181.06 L07181.14
ALFAL07181.06EA 0 SGD
ALFAL07181.06 ALFAL07181.14
2-Bromo-4-nitroanisole ≥98%
2-Bromo-4-nitroanisole

Purity: 98% CAS No.: 5197-28-4 Molecular Formula: C7H6BrNO3 Formula Weight: 232.03 Melting Point: 103-108° Beilstein Reference: 2556372

Formula: C₇H₆BrNO₃
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00041242
CAS Number: 5197-28-4
EINECS: 225-983-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR