2-Bromo-6-fluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

L07511.04
ALFAL07511.04EA 136 SGD
ALFAL07511.04
2-Bromo-6-fluorotoluene ≥98%
2-Bromo-6-fluorotoluene

Purity: 98% CAS No.: 1422-54-4 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 76°(168°F) Refractive Index: 1.5330 Boiling Point: 180° Beilstein Reference: 2433658 MDL No.: MFCD00051438

Formula: C₇H₆BrF
Boiling Pt: 180 °C
Density: 1,53
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051438
CAS Number: 1422-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR