Ethyl-4-nitrophenylglyoxylate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Ethyl 2-(4-nitrophenyl)-2-oxoacetate

L07579.03 L07579.06
ALFAL07579.03EA 121 SGD
ALFAL07579.03 ALFAL07579.06
Ethyl-4-nitrophenylglyoxylate ≥98%
Ethyl 4-nitrophenylglyoxylate

Purity: 98+% CAS No.: 70091-75-7 Molecular Formula: C10H9NO5 Formula Weight: 223.19 Melting Point: 39-43° Boiling Point: 152-157°/0.5mm MDL No.: MFCD00051841

Formula: C₁₀H₉NO₅
MW: 223.19 g/mol
Boiling Pt: 152…157 °C (0.5 mmHg)
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00051841
CAS Number: 70091-75-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR