4-Ethylbenzonitrile 98%

Supplier: Thermo Fisher Scientific

Synonyms: p-Ethylbenzonitrile

L07921.09
ALFAL07921.09EA 213 SGD
ALFAL07921.09
4-Ethylbenzonitrile 98%
4-Ethylbenzonitrile

Purity: 98% CAS No.: 25309-65-3 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 97°(206°F) Refractive Index: 1.5260 Boiling Point: 237° Beilstein Reference: 2040912 MDL No.: MFCD00016383

Formula: C₉H₉N
Boiling Pt: 237 °C
Density: 0,956
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016383
CAS Number: 25309-65-3
EINECS: 246-811-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR