trans-1,2-Bis(diphenylphosphino)ethylene 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (E)-1,2-Bis(diphenylphosphino)ethene

L08508.03
ALFAL08508.03EA 128 SGD
ALFAL08508.03
trans-1,2-Bis(diphenylphosphino)ethylene 98%
trans-1,2-Bis(diphenylphosphino)ethylene

Purity: 98% CAS No.: 983-81-3 Molecular Formula: C26H22P2 Formula Weight: 396.41 Melting Point: 126-130° Beilstein Reference: 757793 MDL No.: MFCD00063219

Formula: C₂₆H₂₂P₂
MW: 396.41 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00063219
CAS Number: 983-81-3
EINECS: 213-570-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR